ENA429360852 Enamine

N-(3-bromophenyl)pyrrolidin-3-amine

  • MDL number MFCD18381822
  • Cas Number : 1379367-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189641
Availability details for
Quantity
  • Availability
  • Add to Cart