ENA429360857 Enamine

3-methylquinolin-5-ol

  • MDL number MFCD18414792
  • Cas Number : 420786-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191015

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191015
Availability details for
Quantity
  • Availability
  • Add to Cart