ENA429360861 Enamine

N-(carbamothioylamino)-2-phenylacetamide

  • MDL number MFCD00758507
  • Cas Number : 29313-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191019
Availability details for
Quantity
  • Availability
  • Add to Cart