ENA429360862 Enamine

1-(5-methyl-1,2-oxazol-3-yl)ethan-1-ol

  • MDL number MFCD16547133
  • Cas Number : 95104-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191020

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191020
Availability details for
Quantity
  • Availability
  • Add to Cart