ENA429360863 Enamine

2-(1-methyl-1H-indazol-5-yl)acetic acid

  • MDL number MFCD11901729
  • Cas Number : 1176749-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189475
Availability details for
Quantity
  • Availability
  • Add to Cart