ENA429360868 Enamine

1-(chloromethyl)-3-(difluoromethyl)benzene

  • MDL number MFCD24225788
  • Cas Number : 1169873-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191095
Availability details for
Quantity
  • Availability
  • Add to Cart