ENA429360869 Enamine

(1S)-1-(3-fluoropyridin-2-yl)ethan-1-ol

  • MDL number MFCD24133488
  • Cas Number : 1016228-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191096

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191096
Availability details for
Quantity
  • Availability
  • Add to Cart