ENA429360873 Enamine

2-cyclopropyl-2-methoxyacetic acid

  • MDL number MFCD21287138
  • Cas Number : 1492471-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191108

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191108
Availability details for
Quantity
  • Availability
  • Add to Cart