ENA429360875 Enamine

(2,2,2-trifluoroethyl)(3,3,3-trifluoropropyl)amine hydrochloride

  • MDL number MFCD28118850
  • Cas Number : 727732-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189648
Availability details for
Quantity
  • Availability
  • Add to Cart