ENA429360879 Enamine

1-(cyclopropylmethyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD21106666
  • Cas Number : 1497772-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191137
Availability details for
Quantity
  • Availability
  • Add to Cart