ENA429360884 Enamine

1-[5-chloro-2-(chloromethyl)phenyl]-1H-1,2,4-triazole

  • MDL number MFCD28125143
  • Cas Number : 1602769-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191152
Availability details for
Quantity
  • Availability
  • Add to Cart