ENA429360885 Enamine

2-[(5-bromopyridin-3-yl)amino]ethan-1-ol

  • MDL number MFCD26801045
  • Cas Number : 1289078-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191154

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191154
Availability details for
Quantity
  • Availability
  • Add to Cart