ENA429360886 Enamine

ethyl 2-[(but-3-en-1-yl)amino]acetate

  • MDL number MFCD13175077
  • Cas Number : 455333-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191160
Availability details for
Quantity
  • Availability
  • Add to Cart