ENA429360889 Enamine

5-iodo-2,4-dimethylbenzoic acid

  • MDL number MFCD12547504
  • Cas Number : 742081-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191186
Availability details for
Quantity
  • Availability
  • Add to Cart