ENA429360892 Enamine

1,3-diethyl 2-(2-nitro-1-phenylethyl)propanedioate

  • MDL number MFCD02941009
  • Cas Number : 22975-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191195

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191195
Availability details for
Quantity
  • Availability
  • Add to Cart