ENA429360893 Enamine

4-chloro-2-(4-chlorophenyl)aniline

  • MDL number MFCD24078169
  • Cas Number : 1101170-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191201

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191201
Availability details for
Quantity
  • Availability
  • Add to Cart