ENA429360896 Enamine

methyl 2-(ethenyloxy)benzoate

  • MDL number MFCD21755511
  • Cas Number : 13661-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189656
Availability details for
Quantity
  • Availability
  • Add to Cart