ENA429360897 Enamine

1-[(4-fluorophenyl)methyl]-3,3-dimethylpiperazine

  • MDL number MFCD16312460
  • Cas Number : 1226207-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191211
Availability details for
Quantity
  • Availability
  • Add to Cart