ENA429360902 Enamine

6,7-difluoro-1-(1-methanesulfonylpropan-2-yl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD20591053
  • Cas Number : 1378723-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189659
Availability details for
Quantity
  • Availability
  • Add to Cart