ENA429360909 Enamine

3-ethyl-5-(2,2,2-trifluoroethyl)-1H-1,2,4-triazole

  • MDL number MFCD14610955
  • Cas Number : 1248135-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191230
Availability details for
Quantity
  • Availability
  • Add to Cart