ENA429360910 Enamine

3-(propan-2-yl)-5-(2,2,2-trifluoroethyl)-1H-1,2,4-triazole

  • MDL number MFCD14627966
  • Cas Number : 1249765-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191232

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191232
Availability details for
Quantity
  • Availability
  • Add to Cart