ENA429360911 Enamine

[1-(2-aminopyrimidin-4-yl)pyrrolidin-2-yl]methanol

  • MDL number MFCD23986828
  • Cas Number : 1533543-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191237

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191237
Availability details for
Quantity
  • Availability
  • Add to Cart