ENA429360912 Enamine

(1R)-1-(3-fluoropyridin-2-yl)ethan-1-ol

  • MDL number MFCD24133489
  • Cas Number : 1568238-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191239
Availability details for
Quantity
  • Availability
  • Add to Cart