ENA429360920 Enamine

4-amino-N-(5-ethyl-1,3,4-thiadiazol-2-yl)benzene-1-sulfonamide

  • MDL number MFCD00057224
  • Cas Number : 94-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-191
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191266
Availability details for
Quantity
  • Availability
  • Add to Cart