ENA429360923 Enamine

N4-(2-phenylethyl)quinazoline-4,7-diamine

  • MDL number MFCD28125156
  • Cas Number : 1462347-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191275
Availability details for
Quantity
  • Availability
  • Add to Cart