ENA429360924 Enamine

methyl 2-(quinolin-2-yl)propanoate

  • MDL number MFCD28125049
  • Cas Number : 97479-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189694
Availability details for
Quantity
  • Availability
  • Add to Cart