ENA429360925 Enamine

4-(piperidin-3-yl)pyridin-2-amine

  • MDL number MFCD17488333
  • Cas Number : 1260649-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191277

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191277
Availability details for
Quantity
  • Availability
  • Add to Cart