ENA429360926 Enamine

5-(3-methoxypropoxy)pyrazine-2-carbonitrile

  • MDL number MFCD20284019
  • Cas Number : 1339933-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191278
Availability details for
Quantity
  • Availability
  • Add to Cart