ENA429360933 Enamine

2-[(naphthalen-2-yl)methyl]oxirane

  • MDL number MFCD22414041
  • Cas Number : 145233-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191300
Availability details for
Quantity
  • Availability
  • Add to Cart