ENA429360934 Enamine

3-methoxy-5-(1H-pyrazol-1-yl)aniline

  • MDL number MFCD23979808
  • Cas Number : 1516412-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191304

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191304
Availability details for
Quantity
  • Availability
  • Add to Cart