ENA429360936 Enamine

3-methoxy-4-(1H-pyrrol-1-yl)aniline hydrochloride

  • MDL number MFCD18783029
  • Cas Number : 1279219-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191312
Availability details for
Quantity
  • Availability
  • Add to Cart