ENA429360940 Enamine

7-bromothieno[3,2-b]pyridine

  • MDL number MFCD18801029
  • Cas Number : 603305-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191318

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-191318
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart