ENA429360943 Enamine

3-[(dimethylamino)methyl]-1H-indole-4-carboxylic acid hydrochloride

  • MDL number MFCD28125162
  • Cas Number : 1326743-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191321
Availability details for
Quantity
  • Availability
  • Add to Cart