ENA429360944 Enamine

1-(thiophene-2-carbonyl)piperidin-4-amine hydrochloride

  • MDL number MFCD09455684
  • Cas Number : 1158299-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191322
Availability details for
Quantity
  • Availability
  • Add to Cart