ENA429360950 Enamine

6-(5-amino-3-methyl-1H-pyrazol-1-yl)-N-methylpyrimidin-4-amine

  • MDL number MFCD24613077
  • Cas Number : 1018473-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189716
Availability details for
Quantity
  • Availability
  • Add to Cart