ENA429360956 Enamine

4-(oxan-4-yloxy)cyclohexane-1-carboxylic acid

  • MDL number MFCD26739547
  • Cas Number : 1558282-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191339
Availability details for
Quantity
  • Availability
  • Add to Cart