ENA429360965 Enamine

4-bromo-1,5-dimethyl-1H-pyrazole-3-sulfonyl chloride

  • MDL number MFCD23777333
  • Cas Number : 1538239-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191369
Availability details for
Quantity
  • Availability
  • Add to Cart