ENA429360967 Enamine

1-ethynyl-3-(methoxymethyl)benzene

  • MDL number MFCD17281936
  • Cas Number : 1339108-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189732

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189732
Availability details for
Quantity
  • Availability
  • Add to Cart