ENA429360968 Enamine

2-chloro-5-phenylaniline

  • MDL number MFCD11933360
  • Cas Number : 57013-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191381
Availability details for
Quantity
  • Availability
  • Add to Cart