ENA429360969 Enamine

3-(5-bromothiophen-2-yl)-6-chloropyridazine

  • MDL number MFCD12819895
  • Cas Number : 58059-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191382
Availability details for
Quantity
  • Availability
  • Add to Cart