ENA429360970 Enamine

{[(2-chloroethyl)sulfanyl]methyl}benzene

  • MDL number MFCD00039372
  • Cas Number : 4332-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189736
Availability details for
Quantity
  • Availability
  • Add to Cart