ENA429360973 Enamine

methyl 3-(4-methylphenyl)-2-sulfanylpropanoate

  • MDL number MFCD25958831
  • Cas Number : 139744-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191386
Availability details for
Quantity
  • Availability
  • Add to Cart