ENA429360975 Enamine

1-cyano-N-[(4-fluorophenyl)methyl]-N-methylmethanesulfonamide

  • MDL number MFCD09041766
  • Cas Number : 926218-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191389
Availability details for
Quantity
  • Availability
  • Add to Cart