ENA429360978 Enamine

2-(1-methyl-1H-pyrazol-5-yl)phenol

  • MDL number MFCD00120278
  • Cas Number : 123532-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-167
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191396
Availability details for
Quantity
  • Availability
  • Add to Cart