ENA429360985 Enamine

4-(3,4-dichlorophenyl)-1,3-oxazol-2-amine

  • MDL number MFCD16667998
  • Cas Number : 1250201-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189742
Availability details for
Quantity
  • Availability
  • Add to Cart