ENA429360986 Enamine

N-(6-aminopyridin-3-yl)benzamide

  • MDL number MFCD17245545
  • Cas Number : 936210-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191424
Availability details for
Quantity
  • Availability
  • Add to Cart