ENA429360988 Enamine

3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine dihydrochloride

  • MDL number MFCD00182087
  • Cas Number : 62002-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191431

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191431
Availability details for
Quantity
  • Availability
  • Add to Cart