ENA429360993 Enamine

2-amino-1-(3-methoxyphenyl)ethan-1-one hydrochloride

  • MDL number MFCD06739758
  • Cas Number : 24037-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191440
Availability details for
Quantity
  • Availability
  • Add to Cart