ENA429360997 Enamine

methyl (3R)-1H,2H,3H,4H,9H-pyrido[3,4-b]indole-3-carboxylate hydrochloride

  • MDL number MFCD23703234
  • Cas Number : 115524-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-253
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-191444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191444
Availability details for
Quantity
  • Availability
  • Add to Cart