ENA429360999 Enamine

2-(4-methylphenyl)-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine dihydrobromide

  • MDL number MFCD19442236
  • Cas Number : 1332530-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191446
Availability details for
Quantity
  • Availability
  • Add to Cart